Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethylene glycol diacetate, 98%

$103.76 - $314.01

Chemical Identifiers

Chemical Name or Material Triethylene glycol diacetate
CAS 111-21-7
Molecular Formula CH3CO2(CH2CH2O)3COCH3
Formula Weight 234.25
MDL Number MFCD00026213
Beilstein 1789453
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL127890B

 
alfa aesar™
L127890B
1000g Each for $314.01

AAL1278930

 
alfa aesar™
L1278930
250g Each for $103.76
Description & Specifications

Specifications

Formula Weight 234.25
Chemical Name or Material Triethylene glycol diacetate
Molecular Formula CH3CO2(CH2CH2O)3COCH3
MDL Number MFCD00026213
CAS 111-21-7
TSCA TSCA
Assay 98%
Beilstein 1789453
Boiling Point 286°C
Melting Point −50°C
Flash Point 163°C (325°F)
RTECS Number YE4950000
Quantity 1000g