missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethylene glycol diacetate, 98%

Manufacturer:  Alfa Aesar™ L1278930

 View more versions of this product

Catalog No. AAL1278930



Specifications

Specifications

Triethylene glycol diacetate
286°C
250g
C10H18O6
1789453
OVOUKWFJRHALDD-UHFFFAOYSA-N
2-[2-(2-acetyloxyethoxy)ethoxy]ethyl acetate
8098
98%
163°C (325°F)
-50°C
111-21-7
MFCD00026213
triethylene glycol diacetate, tdac, triglycol diacetate, triethylene glycol, diacetate, triglycol, diacetate, acetic acid, triethylene glycol diester, triethyleneglycoldiacetate, 2,2'-ethylenedioxy di ethyl acetate, ethanol, 2,2'-ethylenedioxydi-, diacetate, ethanol, 2,2'-1,2-ethanediylbis oxy bis-, diacetate
CC(=O)OCCOCCOCCOC(=O)C
234.248
234.25

Chemical Identifiers

111-21-7
234.248
OVOUKWFJRHALDD-UHFFFAOYSA-N
8098
CC(=O)OCCOCCOCCOC(=O)C
C10H18O6
MFCD00026213
triethylene glycol diacetate, tdac, triglycol diacetate, triethylene glycol, diacetate, triglycol, diacetate, acetic acid, triethylene glycol diester, triethyleneglycoldiacetate, 2,2'-ethylenedioxy di ethyl acetate, ethanol, 2,2'-ethylenedioxydi-, diacetate, ethanol, 2,2'-1,2-ethanediylbis oxy bis-, diacetate
2-[2-(2-acetyloxyethoxy)ethoxy]ethyl acetate
Safety and Handling

Safety and Handling

RTECSNumber : YE4950000

TSCA : TSCA

Documents