missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethylene Glycol Butyl Methyl Ether 98.0+%, TCI America™
Supplier: TCI America T318125ML
Specifications
Triethylene Glycol Butyl Methyl Ether | |
-34°C | |
C11H24O4 | |
25 mL | |
SNAQINZKMQFYFV-UHFFFAOYSA-N | |
2,5,8,11-tetraoxapentadecane | |
18344639 | |
≥98.0% (GC) |
7382-30-1 | |
Colorless | |
MFCD23703159 | |
1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane | |
CCCCOCCOCCOCCOC | |
220.31 | |
220.31 | |
Liquid |
Chemical Identifiers
7382-30-1 | |
220.31 | |
SNAQINZKMQFYFV-UHFFFAOYSA-N | |
18344639 | |
CCCCOCCOCCOCCOC |
C11H24O4 | |
MFCD23703159 | |
1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane | |
2,5,8,11-tetraoxapentadecane |
Safety and Handling
EINECSNumber : (7)-1321
TSCA : No