missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylene Glycol 2-Bromoethyl Methyl Ether, TCI America™

Manufacturer:  TCI America T263425G

 View more versions of this product

Catalog No. T263425G



Specifications

Specifications

Triethylene Glycol 2-Bromoethyl Methyl Ether
25g
C9H19BrO4
YFFFQGXWMHAJPP-UHFFFAOYSA-N
1-[2-[2-(2-bromoethoxy)ethoxy]ethoxy]-2-methoxyethane
19389132
Clear Liquid at 20°C
Yellow
110429-45-3
2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethyl Bromide, 1-Bromo-3,6,9,12-tetraoxatridecane, mPEG4-Bromide
COCCOCCOCCOCCBr
271.151
271.15
≥95.0% (GC)

Chemical Identifiers

110429-45-3
271.151
2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethyl Bromide, 1-Bromo-3,6,9,12-tetraoxatridecane, mPEG4-Bromide
1-[2-[2-(2-bromoethoxy)ethoxy]ethoxy]-2-methoxyethane
C9H19BrO4
YFFFQGXWMHAJPP-UHFFFAOYSA-N
19389132
COCCOCCOCCOCCBr
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents