Please login to your online account to display your discounted pricing

Triethylborate, 97%, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC428651000

 View more versions of this product

Catalog No. AC428651000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 145.99
Quantity 100mL
Packaging AcroSeal™ Glass Bottle
Density 0.8580g/mL
Linear Formula (C2H5O)3B
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
MDL Number MFCD00009073
CAS 150-46-9
Chemical Name or Material Triethyl borate
Assay Percent Range 97%
Molecular Formula C6H15BO3
Synonym Triethoxyborane
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Triethyl borate
Assay Percent Range 97%
Molecular Formula C6H15BO3
Linear Formula (C2H5O)3B
Formula Weight 145.99
MDL Number MFCD00009073
Synonym Triethoxyborane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents