Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethylammonium formate solution 2M (1:1, pH 6.5)

Chemical Identifiers

Chemical Name or Material Triethylammonium formate solution
CAS 585-29-5
Linear Formula (CH3CH2)3NH+ COO-
Formula Weight 147.22
MDL Number MFCD00134943
UN Number UN1760
Beilstein 4150472
Synonym TEAF
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH36155AE

  SDS
alfa aesar™
H36155AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.22
pH 6.5
Density 1.01g/mL
Physical Form Liquid
Beilstein 4150472
Synonym TEAF
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Chemical Name or Material Triethylammonium formate solution
CAS 585-29-5
Health Hazard 2 GHS H Statement
H314-H302-H312-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00134943
UN Number UN1760
Color Colorless
Quantity 100mL
Concentration 2M (1:1)
Linear Formula (CH3CH2)3NH+ COO-
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H302-H312-H332
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.