Please login to your online account to display your discounted pricing

Triethylammonium Acetate, 1 M Solution, Calbiochem™, EMD Millipore

Ready-to-use buffer solution used for the purification of chemically synthesized oligonucleotides by HPLC

$330.00 - $330.00

Chemical Identifiers

Formula Weight 161.2
CAS 5204-74-0
Chemical Name or Material Triethylammonium Acetate
Concentration or Composition (by Analyte or Components) 1M Solution
Molecular Formula C8H19NO2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6257181L

 
emd millipore corporation
6257181L
Each for $330.00
Description & Specifications

Specifications

Formula Weight 161.2
Quantity 1L
Chemical Name or Material Triethylammonium Acetate
Molecular Formula C8H19NO2
CAS 5204-74-0
Concentration or Composition (by Analyte or Components) 1M Solution
Recommended Storage 15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Safety and Handling

15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.