Please login to your online account to display your discounted pricing

Triethylamine (Reagent), Fisher Chemical


Quantity 20L
Packaging Steel Pail

Manufacturer: fisher scientific company  BPO488520

 View more versions of this product

Catalog No. O4885-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Quantity 20L
Purity Grade Reagent Grade
Formula Weight 101.19
Boiling Point 90°C
Melting Point −115°C
DOT Information DOT Class 3, : Flammable Liquid
Density 0.728g/cm³
Physical Form Liquid
Molecular Formula C6H15N
Vapor Pressure 69mbar at 20°C
pH 12.4
Color Colorless
UN Number 1296
Assay Percent Range ≥99%
Synonym N, N-Diethylethanamine
MDL Number MFCD00009051
Additional Information Vapor Pressure: 69mbar at 20°C
ChemAlert Storage Symbol Red
Cleanup Symbol Solvent
Identification Pass Test
Linear Formula (C2H5)3N
CAS 121-44-8
Packaging Steel Pail
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Highly flammable. Harmful by inhalation, in contact with skin and if swallowed. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:3
Instability:0
Chemical Name or Material Triethylamine

Specifications

Chemical Name or Material Triethylamine
CAS 121-44-8
Purity Grade Reagent Grade
Assay ≥99%
Assay Percent Range ≥99%
Molecular Formula C6H15N
Linear Formula (C2H5)3N
Formula Weight 101.19
MDL Number MFCD00009051
Synonym N, N-Diethylethanamine
Accessories
 

Emergency Overview
Highly flammable. Harmful by inhalation, in contact with skin and if swallowed. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents