Learn More
Triethylamine hydrobromide, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 349725000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Triethylamine hydrobromide | |
636-70-4 | |
Authentic | |
Glass bottle | |
(C2H5)3N·HBr | |
500g | |
NRTLTGGGUQIRRT-UHFFFAOYSA-N | |
N,N-diethylethanamine;hydrobromide | |
2734074 | |
99% |
99% | |
>65°C | |
98.5% min. (Argentometry) | |
C6H16BrN | |
MFCD00054287 | |
triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt | |
[Br-].CC[NH+](CC)CC | |
182.11 | |
182.1 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISO
GHS Signal Word: Warning
EINECSNumber : 211-263-8
TSCA : TSCA
RUO â Research Use Only