Please login to your online account to display your discounted pricing

Triethylamine, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle

Manufacturer: avantor performance  W63507

 View more versions of this product

Catalog No. 02004540

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 101.19
Chemical Name or Material Triethylamine
Molecular Formula (C2H5)3N
CAS 121-44-8
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.73kg
Flash Point −11.00°C
Quantity 500mL

Specifications

Chemical Name or Material Triethylamine
CAS 121-44-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula (C2H5)3N
Formula Weight 101.19