missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylamine, 99%, pure, ACROS Organics™

Manufacturer:  Acros Organics AC157911000

 View more versions of this product

Catalog No. AC157911000



Specifications

Specifications

Triethylamine
−11°C
Glass bottle
0.728
0.2% Max. (K.F.)
Undesignated
100mL
99%
(C2H5)3N
01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
101.19
99%
0.7280g/mL
Authentic
1.3990 to 1.4010
69mbar at 20°C
90.0°C
-115.0°C
121-44-8
C6H15N
04, 99
15, 9831
Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
CCN(CC)CC
101.193
CHEBI:35026
Liquid
Pure

Chemical Identifiers

121-44-8
101.193
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC
C6H15N
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine
Safety and Handling

Safety and Handling

GHS P Statement Highly flammable liquid and vapour. Toxic if inhaled. Toxic in contact with skin. Harmful if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse

Danger

SDS Documents