missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylamine, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A12646AP

 View more versions of this product

Catalog No. AAA12646AP



Description

Description

Applications
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizer for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in automotive casting industry and textile industry.

Solubility
Miscible with water, ether and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Triethylamine
500mL
99%
(CH3CH2)3N
UN1296
14,9666
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
101.19
99%
0.7255
121-44-8
C6H15N
MFCD00009051
1843166
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CCN(CC)CC
101.193
CHEBI:35026
Liquid

Chemical Identifiers

121-44-8
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine
C6H15N
MFCD00009051
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H302-H312-H332

GHS P Statement
P210-P260-P261-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIETHYLAMINE

RTECSNumber : YE0175000

TSCA : TSCA

Documents