Learn More
Triethylamine, 99.7%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 219510010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triethylamine | |
121-44-8 | |
0.7280g/mL | |
−11°C | |
99.6% min. (GC) | |
C6H15N | |
(C2H5)3N | |
1 kg | |
01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336 | |
15, 9831 | |
Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride | |
CCN(CC)CC | |
101.19 | |
0.36 mPa.s (20°C) | |
101.19 | |
99.7% | |
Liquid |
99.7% | |
-115.0°C | |
90.0°C | |
Authentic | |
Glass bottle | |
1.3990 to 1.4010 | |
MFCD00009051 | |
04, 99 | |
0.728 | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
N,N-diethylethanamine | |
8471 | |
CHEBI:35026 | |
69mbar at 20°C | |
Extra Pure |
Chemical Identifiers
121-44-8 | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
8471 | |
N,N-diethylethanamine |
MFCD00009051 | |
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
CHEBI:35026 | |
CCN(CC)CC |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
EINECSNumber : 204-469-4
RTECSNumber : YE0175000
TSCA : TSCA
RUO – Research Use Only