missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethylamine, 99+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 12391D5

 View more versions of this product

Catalog No. AA12391D5



Specifications

Specifications

Triethylamine
−11° (12°F)
1.4005
Colorless
3 x 1kg
>99%
(CH3CH2)3N
UN1296
14,9666
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
101.19
≥99%
0.726g/mL
Ammonia-like
89°C to 90°C
-115°C
121-44-8
C6H15N
MFCD00009051
1843166
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CCN(CC)CC
101.193
CHEBI:35026
Liquid

Chemical Identifiers

121-44-8
101.193
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine
C6H15N
MFCD00009051
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIETHYLAMINE

RTECSNumber : YE0175000

TSCA : TSCA

Documents