missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethylaluminum, 25% w/w in hexane, 95%

Manufacturer:  Alfa Aesar™ 89054ND

 View more versions of this product

Catalog No. AA89054ND



Specifications

Specifications

Triethylaluminum
97-93-8
0.832g/mL
C6H15Al
UN3399
VOITXYVAKOUIBA-UHFFFAOYSA-N
triethylalumane
16682930
Liquid
25% w/w in hexane
25g
Air & Moisture Sensitive
MFCD00009015
triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminium, triethyl-aluminium
CC[Al](CC)CC
114.168
114.17
95%

Chemical Identifiers

97-93-8
114.168
VOITXYVAKOUIBA-UHFFFAOYSA-N
16682930
CC[Al](CC)CC
C6H15Al
MFCD00009015
triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminium, triethyl-aluminium
triethylalumane
Safety and Handling

Safety and Handling

GHS H Statement
H224-H250-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE

TSCA : TSCA

SDS Documents