Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethylaluminum, 25% w/w in hexane, 95%

Chemical Identifiers

Chemical Name or Material Triethylaluminum
CAS 97-93-8
Molecular Formula (C2H5)3Al
Formula Weight 114.17
MDL Number MFCD00009015
UN Number UN3399
Synonym TEA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA89054NF

  SDS
alfa aesar™
89054NF
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.17
Physical Form Liquid
Density 0.832g/mL
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Triethylaluminum
Concentration 25% w/w in hexane
Sensitivity Air & Moisture Sensitive
TSCA TSCA
Assay 95%
Molecular Formula (C2H5)3Al
MDL Number MFCD00009015
CAS 97-93-8
Quantity 100g
UN Number UN3399
Synonym TEA
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H250-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H250-H260-H304-H361-H373-H314-H318-H336
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.