Learn More
Triethylaluminium, 0.6M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 381171000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Triethylaluminium | |
97-93-8,142-82-5 | |
0.6880g/mL | |
C6H15Al | |
MFCD00009015 | |
0.688 | |
triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminum, triethyl-aluminum | |
VOITXYVAKOUIBA-UHFFFAOYSA-N | |
triethylalumane | |
16682930 | |
Liquid |
0.6M Solution in Heptane | |
Colorless | |
AcroSeal™ Glass bottle | |
Al(CH2CH3)3 | |
100 mL | |
15, 326 | |
Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons | |
CC[Al](CC)CC | |
114.17 | |
114.17 |
Chemical Identifiers
97-93-8 | |
114.17 | |
VOITXYVAKOUIBA-UHFFFAOYSA-N | |
16682930 | |
CC[Al](CC)CC |
C6H15Al | |
MFCD00009015 | |
triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminum, triethyl-aluminum | |
triethylalumane |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. May be fatal if swallowed and enters airways. Catches fire spontaneously if exposed to air. Highly flammable liquid and vapor. Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container.
GHS Signal Word: Danger
EINECSNumber : 202-619-3
RUO – Research Use Only