missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethyl phosphonoacetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 139700025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for C-C Bond Formation (Olefination)Specifications
Triethyl phosphonoacetate | |
2mg KOH/g max. | |
260.0°C to 262.0°C | |
Authentic | |
Glass bottle | |
1.4300 to 1.4320 | |
2.5kg | |
01,1216; 06,612; 12,527; 13,255 | |
triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate | |
GGUBFICZYGKNTD-UHFFFAOYSA-N | |
ethyl 2-diethoxyphosphorylacetate | |
13345 | |
97% |
867-13-0 | |
1.1300g/mL | |
105°C | |
96% min. (GC) | |
C8H17O5P | |
(C2H5O)2P(O)CH2CO2C2H5 | |
04,I,573 | |
1.13 | |
Solubility in water: insoluble | |
CCOC(=O)CP(=O)(OCC)OCC | |
224.19 | |
224.19 |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 212-757-6
RUO â Research Use Only