missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethyl orthoformate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139660250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
Triethyl orthoformate | |
-61.0°C | |
0.8900g/mL | |
35°C | |
97.5% min. (GC) | |
C7H16O3 | |
CH(OC2H5)3 | |
25 L | |
01,1204; 02,430; 04,527; 05,690; 06,610; 07,385; 11,555; 13,327 | |
15, 6980 | |
Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform, | |
CCOC(OCC)OCC | |
148.2 | |
148.2 | |
Liquid |
122-51-0 | |
Colorless | |
146.0°C | |
Authentic | |
Metal Drum | |
1.3900 to 1.3920 | |
MFCD00009230 | |
02, 20 | |
0.89 | |
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
GKASDNZWUGIAMG-UHFFFAOYSA-N | |
diethoxymethoxyethane | |
31214 | |
98% |
Chemical Identifiers
122-51-0 | |
148.2 | |
GKASDNZWUGIAMG-UHFFFAOYSA-N | |
31214 | |
CCOC(OCC)OCC |
C7H16O3 | |
MFCD00009230 | |
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
diethoxymethoxyethane |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 204-550-4
RUO – Research Use Only