missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl Orthochloroacetate, TCI America™

$210.84 - $670.70

Chemical Identifiers

CAS 51076-95-0
Molecular Formula C8H17ClO3
Molecular Weight (g/mol) 196.671
MDL Number MFCD00142889
InChI Key URFKLQSFBXBOQU-UHFFFAOYSA-N
Synonym 2-chloro-1,1,1-triethoxy-ethane, triethyl orthochloroacetate, 2-chloro-1,1,1-triethoxy ethane, orthochloroacetic acid triethyl ester, ethane, 2-chloro-1,1,1-triethoxy, chlorotriethoxyethane, acmc-209tex, 2-chloro-1, 1,1-triethoxy-ethane, 2-chloranyl-1,1,1-triethoxy-ethane, 2-chloro-1,1,1-triethoxyethane
PubChem CID 305922
IUPAC Name 2-chloro-1,1,1-triethoxyethane
SMILES CCOC(CCl)(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C12881ML View Documents TCI America
C12881ML
1mL Each for $210.84
 
C12885ML View Documents TCI America
C12885ML
5mL Each for $670.70
 
Specifications

Chemical Identifiers

51076-95-0
196.671
URFKLQSFBXBOQU-UHFFFAOYSA-N
305922
CCOC(CCl)(OCC)OCC
C8H17ClO3
MFCD00142889
2-chloro-1,1,1-triethoxy-ethane, triethyl orthochloroacetate, 2-chloro-1,1,1-triethoxy ethane, orthochloroacetic acid triethyl ester, ethane, 2-chloro-1,1,1-triethoxy, chlorotriethoxyethane, acmc-209tex, 2-chloro-1, 1,1-triethoxy-ethane, 2-chloranyl-1,1,1-triethoxy-ethane, 2-chloro-1,1,1-triethoxyethane
2-chloro-1,1,1-triethoxyethane

Specifications

91°C
1mL
C8H17ClO3
2-chloro-1,1,1-triethoxy-ethane, triethyl orthochloroacetate, 2-chloro-1,1,1-triethoxy ethane, orthochloroacetic acid triethyl ester, ethane, 2-chloro-1,1,1-triethoxy, chlorotriethoxyethane, acmc-209tex, 2-chloro-1, 1,1-triethoxy-ethane, 2-chloranyl-1,1,1-triethoxy-ethane, 2-chloro-1,1,1-triethoxyethane
CCOC(CCl)(OCC)OCC
196.671
196.67
≥95.0% (GC)
Colorless
51076-95-0
MFCD00142889
URFKLQSFBXBOQU-UHFFFAOYSA-N
2-chloro-1,1,1-triethoxyethane
305922
Clear Liquid at 20°C
Triethyl Orthochloroacetate
Safety and Handling

Safety and Handling

TSCA : No