missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl Orthobutyrate, TCI America™

Manufacturer:  TCI America O017325ML

 View more versions of this product

Catalog No. O017325ML



Specifications

Specifications

Triethyl Orthobutyrate
Colorless
24964-76-9
MFCD00059382
Ethyl Orthobutyrate, Orthobutyric Acid Triethyl Ester, 1,1,1-Triethoxybutane
CCCC(OCC)(OCC)OCC
190.283
190.28
≥90.0% (GC)
63°C
25mL
C10H22O3
3272
KOPMZTKUZCNGFY-UHFFFAOYSA-N
1,1,1-triethoxybutane
371779
Clear Liquid at 20°C

Chemical Identifiers

24964-76-9
190.283
KOPMZTKUZCNGFY-UHFFFAOYSA-N
371779
CCCC(OCC)(OCC)OCC
C10H22O3
MFCD00059382
Ethyl Orthobutyrate, Orthobutyric Acid Triethyl Ester, 1,1,1-Triethoxybutane
1,1,1-triethoxybutane
Safety and Handling

Safety and Handling

TSCA : No

Documents