missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethyl Methanetricarboxylate, TCI America™

Manufacturer:  TCI America T148125G

 View more versions of this product

Catalog No. T148125G



Specifications

Specifications

Triethyl Methanetricarboxylate
White
6279-86-3
MFCD00009150
AGZPNUZBDCYTBB-UHFFFAOYSA-N
triethyl methanetricarboxylate
80471
Crystal-Lump at 20°C
253°C
25g
C10H16O6
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
CCOC(=O)C(C(=O)OCC)C(=O)OCC
232.232
232.23
≥97.0% (GC)

Chemical Identifiers

6279-86-3
232.232
AGZPNUZBDCYTBB-UHFFFAOYSA-N
80471
CCOC(=O)C(C(=O)OCC)C(=O)OCC
C10H16O6
MFCD00009150
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
triethyl methanetricarboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents