missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethyl methanetricarboxylate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 139631000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Triethyl methanetricarboxylate | |
1.1000g/mL | |
>110°C | |
98% | |
C10H16O6 | |
1.4240 to 1.4260 | |
02,81 | |
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane | |
AGZPNUZBDCYTBB-UHFFFAOYSA-N | |
triethyl methanetricarboxylate | |
80471 | |
98% |
6279-86-3 | |
253.0°C | |
Authentic | |
Glass bottle | |
CH(CO2C2H5)3 | |
100mL | |
1.1 | |
Solubility in water: immiscible | |
CCOC(=O)C(C(=O)OCC)C(=O)OCC | |
232.23 | |
232.23 |
Safety and Handling
EINECSNumber : 228-477-2
RUO â Research Use Only