Learn More
Triethyl methanetricarboxylate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0835614
Description
Methanetricarboxylates have been used as blocked malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethanetricarboxylates have been used as ″blocked″ malonic esters, restricting reaction with alkyl halides to monoalkylation. The unwanted carboxyl group can be removed by treatment with Na alkoxide, LDA or BCl3. Provides a useful 2-carbon chain extension method that adds to Michael acceptors under phase-transfer conditions. Triethyl Methanetricarboxylate is used in the synthesis of novel inhibitors of Hsp90. It is also used in the preparation of novel dihydroquinoline-3-carboxylic acids that function as HIV-1 integrase inhibitors.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Triethyl methanetricarboxylate | |
26°C to 29°C | |
253°C | |
C10H16O6 | |
MFCD00009150 | |
1793645 | |
Insoluble in water. | |
CCOC(=O)C(C(=O)OCC)C(=O)OCC | |
232.232 | |
232.23 |
6279-86-3 | |
1.095 | |
>110°C (230°F) | |
1.4255 | |
25 g | |
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane | |
AGZPNUZBDCYTBB-UHFFFAOYSA-N | |
triethyl methanetricarboxylate | |
80471 | |
98% |
Chemical Identifiers
6279-86-3 | |
232.232 | |
AGZPNUZBDCYTBB-UHFFFAOYSA-N | |
80471 | |
CCOC(=O)C(C(=O)OCC)C(=O)OCC |
C10H16O6 | |
MFCD00009150 | |
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane | |
triethyl methanetricarboxylate |
Safety and Handling
EINECSNumber : 228-477-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only