missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyl citrate, 99%

Manufacturer:  Alfa Aesar™ L1263930

 View more versions of this product

Catalog No. AAL1263930



Description

Description

Applications
Triethyl citrate acts as a plasticizer for polyvinylchloride and plastics. It is also useful in pharmaceutical coatings. It is employed as a dye entrapment for the preparation of emission membrane filters for contact fluorescence imaging. It is also employed in fabrication of green nanocomposites from cellulose acetate. It finds application as a pseudo-emulsifier in e-cigarette juices. Further, it functions like Lecithin and utilized in food products. In addition to this, it acts as a carrier reagent and sequestrant.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Triethyl citrate
126°C to 127°C (1mmHg)
250g
C12H20O7
1801199
triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O
276.285
276.29
176°C (348°F)
-55°C
77-93-0
MFCD00009201
14,2326
DOOTYTYQINUNNV-UHFFFAOYSA-N
triethyl 2-hydroxypropane-1,2,3-tricarboxylate
6506
99%

Chemical Identifiers

77-93-0
276.285
DOOTYTYQINUNNV-UHFFFAOYSA-N
6506
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O
C12H20O7
MFCD00009201
triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester
triethyl 2-hydroxypropane-1,2,3-tricarboxylate
Safety and Handling

Safety and Handling

RTECSNumber : GE8050000

TSCA : TSCA

Documents