Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyl borate, 98+%

Chemical Identifiers

Chemical Name or Material Triethyl borate
CAS 150-46-9
Formula Weight 146.00g/mol
MDL Number MFCD00009073
UN Number UN1176
Synonym Triethoxyborane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2297218

 
alfa aesar™
22972-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.858g/mL
Odor Mild
Quantity 50g
Refractive Index 1.381
Boiling Point 117° to 118°C
Melting Point −85°C
Color Colorless
Flash Point 11° (52°F)
Quantity 50g
Chemical Name or Material Triethyl borate
CAS 150-46-9
Physical Form Liquid
Assay ≥98%
Formula Weight 146.00g/mol
MDL Number MFCD00009073
UN Number UN1176
Synonym Triethoxyborane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL BORATE
RTECS Number ED5075000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.