missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyl 4-phosphonobutyrate, 97%

Manufacturer:  Alfa Aesar™ L1048809

 View more versions of this product

Catalog No. AAL1048809



Specifications

Specifications

Triethyl 4-phosphonobutyrate
10g
C10H21O5P
1791802
FYESTJSRASJZOY-UHFFFAOYSA-N
ethyl 4-diethoxyphosphorylbutanoate
281201
97%
117°C to 119°C (0.4mmHg)
2327-69-7
MFCD00015168
triethyl 4-phosphonobutyrate, ethyl 4-diethoxyphosphoryl butanoate, triethyl4-phosphonobutyrate, butanoic acid, 4-diethoxyphosphinyl-, ethyl ester, triethyl-4-phosphono butyrate, butyric acid, 4-diethylphosphono-, ethyl ester, triethyl 4-phosphonobutanoate, 4-phosphonobutanoic acid triethyl ester, 4-diethoxyphosphinyl butyric acid ethyl ester, butanoic acid,4-diethoxyphosphinyl-,ethyl ester
CCOC(=O)CCCP(=O)(OCC)OCC
252.247
252.25

Chemical Identifiers

2327-69-7
252.247
FYESTJSRASJZOY-UHFFFAOYSA-N
281201
CCOC(=O)CCCP(=O)(OCC)OCC
C10H21O5P
MFCD00015168
triethyl 4-phosphonobutyrate, ethyl 4-diethoxyphosphoryl butanoate, triethyl4-phosphonobutyrate, butanoic acid, 4-diethoxyphosphinyl-, ethyl ester, triethyl-4-phosphono butyrate, butyric acid, 4-diethylphosphono-, ethyl ester, triethyl 4-phosphonobutanoate, 4-phosphonobutanoic acid triethyl ester, 4-diethoxyphosphinyl butyric acid ethyl ester, butanoic acid,4-diethoxyphosphinyl-,ethyl ester
ethyl 4-diethoxyphosphorylbutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents