missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethyl 3-phosphonopropionate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1019018
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triethyl 3-phosphonopropionate | |
1.094 | |
>110°C (230°F) | |
1.433 | |
50 g | |
ethyl 3-diethoxyphosphoryl propanoate, triethyl 3-phosphonopropionate, 3-diethylphosphonopropionic acid ethyl ester, triethyl 3-phosphonopropanoate, ethyl 3-diethoxyphosphinyl propionate, ethyl 3-diethoxycarbonyl propanoate, propanoic acid, 3-diethoxyphosphinyl-, ethyl ester, ethyl 3-diethylphosphonopropionate, ethyl 3-diethylphosphono-propionate, ethyl 3-diethylphosphono propanoate | |
CCOC(=O)CCP(=O)(OCC)OCC | |
238.22 | |
238.22 |
3699-67-0 | |
127°C to 129°C (2 mmHg) | |
C9H19O5P | |
MFCD00015167 | |
1790966 | |
JMJWCUOIOKBVNQ-UHFFFAOYSA-N | |
ethyl 3-diethoxyphosphorylpropanoate | |
281204 | |
98% |
Chemical Identifiers
3699-67-0 | |
238.22 | |
JMJWCUOIOKBVNQ-UHFFFAOYSA-N | |
281204 | |
CCOC(=O)CCP(=O)(OCC)OCC |
C9H19O5P | |
MFCD00015167 | |
ethyl 3-diethoxyphosphoryl propanoate, triethyl 3-phosphonopropionate, 3-diethylphosphonopropionic acid ethyl ester, triethyl 3-phosphonopropanoate, ethyl 3-diethoxyphosphinyl propionate, ethyl 3-diethoxycarbonyl propanoate, propanoic acid, 3-diethoxyphosphinyl-, ethyl ester, ethyl 3-diethylphosphonopropionate, ethyl 3-diethylphosphono-propionate, ethyl 3-diethylphosphono propanoate | |
ethyl 3-diethoxyphosphorylpropanoate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only