missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyl 2-phosphonopentanoate, 98%

Manufacturer:  Alfa Aesar™ 3041304

 View more versions of this product

Catalog No. AA3041304



Specifications

Specifications

Triethyl 2-phosphonopentanoate
35051-49-1
C11H23O5P
MFCD00015163
CCCC(C(=O)OCC)P(=O)(OCC)OCC
266.274
266.27
98°C (2mmHg)
2g
98%
(C2H5O)2P(O)[CH(C3H7)CO2C2H5]
BUPVIVDUPRDDFI-UHFFFAOYSA-N
ethyl 2-diethoxyphosphorylpentanoate
98244
Liquid
Colorless

Chemical Identifiers

35051-49-1
266.274
BUPVIVDUPRDDFI-UHFFFAOYSA-N
ethyl 2-diethoxyphosphorylpentanoate
C11H23O5P
MFCD00015163
98244
CCCC(C(=O)OCC)P(=O)(OCC)OCC
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

SDS Documents