Please login to your online account to display your discounted pricing

Triethyl 2-fluoro-2-phosphonoacetate, 96%, Acros Organics

Chemical Identifiers

Chemical Name or Material Triethyl 2-fluoro-2-phosphonoacetate
Assay Percent Range 96%
Molecular Formula C8H16FO5P
Formula Weight 242.19
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC375560050

 
thermo fisher scientific
AC375560050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC375560010

 
thermo fisher scientific
AC375560010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.19
Quantity 5g
Chemical Name or Material Triethyl 2-fluoro-2-phosphonoacetate
Assay Percent Range 96%
Molecular Formula C8H16FO5P
CAS 2356-16-3
Packaging Glass bottle

For C-C Bond Formation (Olefination)