Please login to your online account to display your discounted pricing

Triethyl 2-fluoro-2-phosphonoacetate, 96%, Acros Organics


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC375560050

 View more versions of this product

Catalog No. AC375560050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 242.19
Quantity 5g
Chemical Name or Material Triethyl 2-fluoro-2-phosphonoacetate
Assay Percent Range 96%
Molecular Formula C8H16FO5P
CAS 2356-16-3
Packaging Glass bottle

Specifications

Chemical Name or Material Triethyl 2-fluoro-2-phosphonoacetate
Assay Percent Range 96%
Molecular Formula C8H16FO5P
Formula Weight 242.19