Learn More
Triethyl 2-fluoro-2-phosphonoacetate, 96%, Thermo Scientific Chemicals
$85.68 - $785.66
Chemical Identifiers
CAS | 2356-16-3 |
---|---|
Molecular Formula | C8H16FO5P |
Molecular Weight (g/mol) | 242.183 |
MDL Number | MFCD00134400 |
InChI Key | FVPISMANESAJQZ-UHFFFAOYSA-N |
Synonym | triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester |
PubChem CID | 2734865 |
IUPAC Name | ethyl 2-diethoxyphosphoryl-2-fluoroacetate |
SMILES | CCOC(=O)C(F)P(=O)(OCC)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1791303
|
Thermo Scientific Chemicals
L1791303 |
1 g |
N/A
|
|
|||||
AAL1791306
|
Thermo Scientific Chemicals
L1791306 |
5 g |
N/A
|
|
|||||
AAL1791314
|
Thermo Scientific Chemicals
L1791314 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2356-16-3 | |
242.183 | |
FVPISMANESAJQZ-UHFFFAOYSA-N | |
2734865 | |
CCOC(=O)C(F)P(=O)(OCC)OCC |
C8H16FO5P | |
MFCD00134400 | |
triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester | |
ethyl 2-diethoxyphosphoryl-2-fluoroacetate |
Specifications
2356-16-3 | |
75°C (0.01 mmHg) | |
C8H16FO5P | |
MFCD00134400 | |
1912161 | |
FVPISMANESAJQZ-UHFFFAOYSA-N | |
ethyl 2-diethoxyphosphoryl-2-fluoroacetate | |
2734865 | |
96% |
1.194 | |
73°C (163°F) | |
1.425 | |
1 g | |
triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester | |
CCOC(=O)C(F)P(=O)(OCC)OCC | |
242.183 | |
242.19 | |
Triethyl 2-fluoro-2-phosphonoacetate |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only