Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyl 1,1,2-ethanetricarboxylate, 99%

$43.84 - $119.49

Chemical Identifiers

Chemical Name or Material Triethyl 1,1,2-ethanetricarboxylate
CAS 7459-46-3
Molecular Formula C11H18O6
Formula Weight 246.26
MDL Number MFCD00009154
Beilstein 1796294
Synonym 1,1,2-Ethanetricarboxylic acid triethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1201406

  SDS
alfa aesar™
L1201406
5g Each for $43.84

AAL1201414

  SDS
alfa aesar™
L1201414
25g Each for $119.49
Description & Specifications

Specifications

Formula Weight 246.26
MDL Number MFCD00009154
Assay 99%
Molecular Formula C11H18O6
Synonym 1,1,2-Ethanetricarboxylic acid triethyl ester
Quantity 5g
Boiling Point 104° to 105°C (1mmHg)
Beilstein 1796294
Flash Point >110°C (230°F)
Chemical Name or Material Triethyl 1,1,2-ethanetricarboxylate
CAS 7459-46-3