missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triethyl 1,1,2-ethanetricarboxylate, 99%

Manufacturer:  Alfa Aesar™ L1201406

 View more versions of this product

Catalog No. AAL1201406



Specifications

Specifications

Triethyl 1,1,2-ethanetricarboxylate
104°C to 105°C (1mmHg)
7459-46-3
MFCD00009154
triethyl 1,1,2-ethanetricarboxylate, 1,1,2-ethanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-succinic acid diethyl ester, triethyl ethane tricarboxylate, 1,1,2-triethyl ethane-1,1,2-tricarboxylate, triethyl ethane-1,2,2-tricarboxylate, ethane-1,1,2-tricarboxylic acid triethyl ester, ethane-1,1,2-tricarboxylic acid, triethyl ester, diethyl 2-ethoxycarbonyl butane-1,4-dioate, acmc-209ovd
CCOC(=O)CC(C(=O)OCC)C(=O)OCC
246.259
246.26
>110°C (230°F)
5g
C11H18O6
1796294
TVWZLLYAJDSSCJ-UHFFFAOYSA-N
triethyl ethane-1,1,2-tricarboxylate
81961
99%

Chemical Identifiers

7459-46-3
246.259
TVWZLLYAJDSSCJ-UHFFFAOYSA-N
81961
CCOC(=O)CC(C(=O)OCC)C(=O)OCC
C11H18O6
MFCD00009154
triethyl 1,1,2-ethanetricarboxylate, 1,1,2-ethanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-succinic acid diethyl ester, triethyl ethane tricarboxylate, 1,1,2-triethyl ethane-1,1,2-tricarboxylate, triethyl ethane-1,2,2-tricarboxylate, ethane-1,1,2-tricarboxylic acid triethyl ester, ethane-1,1,2-tricarboxylic acid, triethyl ester, diethyl 2-ethoxycarbonyl butane-1,4-dioate, acmc-209ovd
triethyl ethane-1,1,2-tricarboxylate
Documents