missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethoxy(pentafluorophenyl)silane, TCI America™

Manufacturer:  TCI America T31341G

 View more versions of this product

Catalog No. T31341G



Specifications

Specifications

Triethoxy(pentafluorophenyl)silane
Yellow
20083-34-5
MFCD03411257
CCO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC
330.326
330.33
≥95.0% (GC)
238°C
1g
C12H15F5O3Si
QALDFNLNVLQDSP-UHFFFAOYSA-N
triethoxy-(2,3,4,5,6-pentafluorophenyl)silane
2782625
Clear Liquid at 20°C

Chemical Identifiers

20083-34-5
330.326
QALDFNLNVLQDSP-UHFFFAOYSA-N
triethoxy-(2,3,4,5,6-pentafluorophenyl)silane
C12H15F5O3Si
MFCD03411257
2782625
CCO[Si](C1=C(C(=C(C(=C1F)F)F)F)F)(OCC)OCC
Safety and Handling

Safety and Handling

TSCA : No

Documents