missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 97.0+%, TCI America™
Supplier: TCI America T28605G
Specifications
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane | |
Colorless | |
5 g | |
NYIKUOULKCEZDO-UHFFFAOYSA-N | |
triethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane | |
22600765 | |
≥97.0% (GC) |
102390-98-7 | |
C12H19F9O3Si | |
1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane | |
CCO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC | |
410.352 | |
410.35 | |
Liquid |
Chemical Identifiers
102390-98-7 | |
410.352 | |
1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane | |
triethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane |
C12H19F9O3Si | |
NYIKUOULKCEZDO-UHFFFAOYSA-N | |
22600765 | |
CCO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC |
Safety and Handling
TSCA : No