missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane 97.0+%, TCI America™
Supplier: TCI America T286025G
Specifications
| Triethoxy(1H,1H,2H,2H-nonafluorohexyl)silane | |
| Colorless | |
| 25 g | |
| NYIKUOULKCEZDO-UHFFFAOYSA-N | |
| triethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane | |
| 22600765 | |
| ≥97.0% (GC) |
| 102390-98-7 | |
| C12H19F9O3Si | |
| 1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane | |
| CCO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC | |
| 410.352 | |
| 410.35 | |
| Liquid |
Chemical Identifiers
| 102390-98-7 | |
| 410.352 | |
| 1H,1H,2H,2H-Nonafluorohexyltriethoxysilane, Triethoxy(1H,1H,2H,2H-perfluorohexyl)silane | |
| triethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane |
| C12H19F9O3Si | |
| NYIKUOULKCEZDO-UHFFFAOYSA-N | |
| 22600765 | |
| CCO[Si](CCC(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC |
Safety and Handling
TSCA : No