missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane 98.0+%, TCI America™

Manufacturer:  Tci America T28765G

Catalog No. T28765G



Specifications

Specifications

Triethoxy-1H,1H,2H,2H-heptadecafluorodecylsilane
Colorless
101947-16-4
MFCD00042334
MLXDKRSDUJLNAB-UHFFFAOYSA-N
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
145374
Clear Liquid at 20°C
230°C
5g
C16H19F17O3Si
1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan
CCO[Si](CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC
610.383
610.38
≥98.0% (GC)

Chemical Identifiers

101947-16-4
610.383
MLXDKRSDUJLNAB-UHFFFAOYSA-N
145374
CCO[Si](CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OCC)OCC
C16H19F17O3Si
MFCD00042334
1h,1h,2h,2h-perfluorodec-1-yl tris ethoxy silan
triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)silane
Safety and Handling

Safety and Handling

TSCA : No

Documents