Please login to your online account to display your discounted pricing

Triethanolamine salicylate, 99%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  269842500

 View more versions of this product

Catalog No. AC269842500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Formula Weight 287.3
Quantity 250g
Color Pale yellow to amber
Assay 98.5 to 101.5%
pH 6 to 7 (5% soln.)
Merck Index 15, 9830
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C6H15NO3·C7H6O3
MDL Number MFCD00082325
CAS 2174-16-5
Assay Percent Range 99%
Name Note 99%
Chemical Name or Material Triethanolamine salicylate
Linear Formula OHC6H4CO2H·N(CH2CH2OH)3

Specifications

Chemical Name or Material Triethanolamine salicylate
Name Note 99%
CAS 2174-16-5
Assay 98.5 to 101.5%
Assay Percent Range 99%
Molecular Formula C6H15NO3·C7H6O3
Linear Formula OHC6H4CO2H·N(CH2CH2OH)3
Formula Weight 287.3
MDL Number MFCD00082325