Please login to your online account to display your discounted pricing

Triethanolamine salicylate, 99%, ACROS Organics™

$107.18 - $107.18

Chemical Identifiers

Chemical Name or Material Triethanolamine salicylate
Name Note 99%
CAS 2174-16-5
Assay 98.5 to 101.5%
Assay Percent Range 99%
Molecular Formula C6H15NO3·C7H6O3
Linear Formula OHC6H4CO2H·N(CH2CH2OH)3
Formula Weight 287.3
MDL Number MFCD00082325
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC269842500

  SDS
thermo fisher scientific
269842500
Glass bottle 250g Each for $107.18
Description & Specifications

Specifications

Physical Form Crystalline Solid
Formula Weight 287.3
Quantity 250g
Color Pale yellow to amber
Assay 98.5 to 101.5%
pH 6 to 7 (5% soln.)
Merck Index 15, 9830
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C6H15NO3·C7H6O3
MDL Number MFCD00082325
CAS 2174-16-5
Assay Percent Range 99%
Name Note 99%
Chemical Name or Material Triethanolamine salicylate
Linear Formula OHC6H4CO2H·N(CH2CH2OH)3