Please login to your online account to display your discounted pricing

Triethanolamine (NF), Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  T350500

 View more versions of this product

Catalog No. T350-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Formula Weight 149.14
Boiling Point 360°C
Melting Point 21°C
Vapor Pressure <0.01 mmHg at 20°C
Quantity 500mL
Purity Grade NF
Assay 99.0 to 107.4%
Density 1.125g/cm³
Molecular Formula C6H15NO3
Decomposition Information >325°C
pH 10.5
Color Light Yellow
Water 0.5% max.
MDL Number MFCD00002855
ChemAlert Storage Symbol Gray
Synonym Tri-beta-hydroxy Ethylamine
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0
Assay Percent Range 99.0 to 107.4 %
Identification Pass Test
Additional Information Vapor Pressure: <0.01mmHg at 20°C
Packaging Glass Bottle
Residual Solvents Meets Requirements (NF)
Specific Gravity 1.120 to 1.128 (at 25°C)
Ignition Residue 0.05% max.
Refractive Index 1.481 to 1.486 (at 20°C)
Chemical Name or Material Triethanolamine
Linear Formula N(CH2CH2OH)3
CAS 102-71-6

Specifications

Chemical Name or Material Triethanolamine
CAS 102-71-6
Purity Grade NF
Assay 99.0 to 107.4%
Assay Percent Range 99.0 to 107.4 %
Molecular Formula C6H15NO3
Linear Formula N(CH2CH2OH)3
Formula Weight 149.14
MDL Number MFCD00002855
Synonym Tri-beta-hydroxy Ethylamine
Accessories
 

Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!