Please login to your online account to display your discounted pricing

Triethanolamine (NF), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  T3504

 View more versions of this product

Catalog No. T350-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Viscous Liquid
Formula Weight 149.14
Melting Point 21°C
Boiling Point 360°C
Vapor Pressure <0.01 mmHg at 20°C
Quantity 4L
Assay 99.0 to 107.4%
Molecular Formula C6H15NO3
Purity Grade NF
Density 1.125g/cm³
pH 10.5
Color Light Yellow
Decomposition Information >325°C
Water 0.5% max.
Synonym Tri-beta-hydroxy Ethylamine
Residual Solvents Meets Requirements (NF)
Refractive Index 1.481 to 1.486 (at 20°C)
MDL Number MFCD00002855
Ignition Residue 0.05% max.
Additional Information Vapor Pressure: <0.01mmHg at 20°C
Identification Pass Test
Assay Percent Range 99.0 to 107.4 %
Packaging Glass Bottle
Specific Gravity 1.120 to 1.128 (at 25°C)
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0
Chemical Name or Material Triethanolamine
ChemAlert Storage Symbol Gray
CAS 102-71-6
Linear Formula N(CH2CH2OH)3

Specifications

Chemical Name or Material Triethanolamine
CAS 102-71-6
Purity Grade NF
Assay 99.0 to 107.4%
Assay Percent Range 99.0 to 107.4 %
Molecular Formula C6H15NO3
Linear Formula N(CH2CH2OH)3
Formula Weight 149.14
MDL Number MFCD00002855
Synonym Tri-beta-hydroxy Ethylamine
Accessories

Accessories for Catalog Number: T3504

Item Mfr. No. Item Description Price Per Unit Quantity  
03-439
Eagle™ Thermoplastics™ Safety Bottle Carriers
fisher scientific
B102
Safety bottle carriers are molded-in handle Each for $217.50
Accessories
 

Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!

SDS & Safety Documents