missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triethanolamine (NF), Fisher Chemical™
$265.86 - $1,428.71

Chemical Identifiers
CAS | 102-71-6 |
---|---|
Molecular Formula | C6H15NO3 |
Molecular Weight (g/mol) | 149.19 |
MDL Number | MFCD00002855 |
InChI Key | GSEJCLTVZPLZKY-UHFFFAOYSA-N |
Synonym | 2,2',2-nitrilotriethanol, daltogen, nitrilotriethanol, sterolamide, thiofaco t-35, triethanolamine, triethylolamine, trihydroxytriethylamine, tris 2-hydroxyethyl amine, trolamine |
PubChem CID | 7618 |
ChEBI | CHEBI:28621 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]ethanol |
SMILES | C(CO)N(CCO)CCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
T350-500
|
Fisher Chemical
T350500 |
500mL | Glass Bottle |
Each for $265.86
Case of 6 Each for $1,595.16 |
|
T350-4
|
Fisher Chemical
T3504 |
4L | Glass Bottle |
Each for $1,428.71
Case of 4 Each for $5,714.84 |
|
Chemical Identifiers
102-71-6 | |
149.19 | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
7618 | |
2-[bis(2-hydroxyethyl)amino]ethanol |
C6H15NO3 | |
MFCD00002855 | |
2,2',2-nitrilotriethanol, daltogen, nitrilotriethanol, sterolamide, thiofaco t-35, triethanolamine, triethylolamine, trihydroxytriethylamine, tris 2-hydroxyethyl amine, trolamine | |
CHEBI:28621 | |
C(CO)N(CCO)CCO |
Specifications
>325°C | |
Pass Test | |
Glass Bottle | |
Meets Requirements (NF) | |
<0.01mmHg at 20°C | |
360°C | |
21°C | |
10.5 | |
99.0 to 107.4 % | |
N(CH2CH2OH)3 | |
2,2',2-nitrilotriethanol, daltogen, nitrilotriethanol, sterolamide, thiofaco t-35, triethanolamine, triethylolamine, trihydroxytriethylamine, tris 2-hydroxyethyl amine, trolamine | |
C(CO)N(CCO)CCO | |
149.19 | |
CHEBI:28621 | |
Viscous Liquid | |
NF |
1.125g/cm³ | |
0.05% max. | |
1.481 to 1.486 (at 20°C) | |
1.120 to 1.128 (at 25°C) | |
0.5% max. | |
Yellow | |
500mL | |
102-71-6 | |
C6H15NO3 | |
MFCD00002855 | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]ethanol | |
7618 | |
149.14 | |
99.0 to 107.4% | |
Triethanolamine |
Safety and Handling

WARNING!
Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.
NFPA
Health:2
Flammability:1
Instability:0