Please login to your online account to display your discounted pricing

Triethanolamine (Certified), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  T4074

 View more versions of this product

Catalog No. T407-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Purity Grade Certified Grade
Physical Form Viscous Liquid
Formula Weight 149.14
Boiling Point 360°C
Melting Point 21°C
Vapor Pressure <0.01 mmHg at 20°C
Quantity 4L
Density 1.125g/cm³
Molecular Formula C6H15NO3
Decomposition Information >325°C
pH 10.5
Color Light Yellow
Assay Percent Range ≥99 %
Additional Information Vapor Pressure: <0.01mmHg at 20°C
Synonym Tri-β-hydroxy Ethylamine
Water 0.2% max.
Iron (Fe) 0.001% max.
MDL Number MFCD00002855
ChemAlert Storage Symbol Gray
Packaging Glass Bottle
Identification Pass Test
Linear Formula N(CH2CH2OH)3
Health Hazard 2 WARNING!
Health Hazard 3 Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0
Chemical Name or Material Triethanolamine
CAS 102-71-6

Specifications

Chemical Name or Material Triethanolamine
CAS 102-71-6
Purity Grade Certified Grade
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula C6H15NO3
Linear Formula N(CH2CH2OH)3
Formula Weight 149.14
MDL Number MFCD00002855
Synonym Tri-β-hydroxy Ethylamine
Accessories

Accessories for Catalog Number: T4074

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Emergency Overview
Irritating to eyes. May cause an allergic skin reaction. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Obtain medical attention. Do not induce vomiting.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!