Please login to your online account to display your discounted pricing

Triethanolamine, +99%, ACROS Organics™


Quantity 2.5L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  421630025

 View more versions of this product

Catalog No. AC421630025

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 149.19
Physical Form Oily Liquid
Quantity 2.5L
Color Clear colorless to slightly yellow
Fieser 01,1196
Merck Index 15, 9830
Packaging Glass Bottle
Density 1.1245g/mL
Synonym 2, 2';, 2';';-Nitrilotriethanol
Assay Percent Range 99+%
Molecular Formula C6H15NO3
Infrared Spectrum Authentic
Refractive Index 1.4830 to 1.4860 (20°C, 589nm)
Chemical Name or Material Triethanolamine
Linear Formula (HOCH2CH2)3N
CAS 102-71-6
MDL Number MFCD00002855
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Triethanolamine
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H15NO3
Linear Formula (HOCH2CH2)3N
Formula Weight 149.19
MDL Number MFCD00002855
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents