Learn More
Triethanolamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 139560025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triethanolamine | |
102-71-6 | |
21.0°C | |
360.0°C | |
Authentic | |
Glass bottle | |
1.4840 to 1.4860 | |
MFCD00002855 | |
01,1196 | |
15, 9830 | |
Solubility in water: freely soluble. Other solubilities: miscible with alcohol,methanol and acetone,soluble in chloroform,slichtly soluble in benzene and ether,soluble 4.2% in benzene; 1.6% in ether; 0.4% in,carbon tetrachloride and < 0.1% in n-he | |
C(CO)N(CCO)CCO | |
149.19 | |
600 mPa.s (25°C) | |
149.19 | |
97% |
97% | |
>325°C | |
1.1245g/mL | |
190°C | |
96% min. (GC) | |
C6H15NO3 | |
(HOCH2CH2)3N | |
2.5 L | |
1.1245 | |
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35 | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]ethanol | |
7618 | |
CHEBI:28621 | |
<0.01mmHg at 20°C | |
Oily Liquid |
Chemical Identifiers
102-71-6 | |
MFCD00002855 | |
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35 | |
CHEBI:28621 |
149.19 | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
7618 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
HYGROSCOPIC
EINECSNumber : 203-049-8
RTECSNumber : KL9275000
TSCA : TSCA
RUO – Research Use Only