missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tricyclohexyltin chloride, 97%

Manufacturer:  Alfa Aesar™ H5549306

 View more versions of this product

Catalog No. AAH5549306



Description

Description

Applications
Tricyclohexyltin chloride is used in the preparation of dicyclohexyl disulfide. It is also used in the synthesis of organotin derivatives of 2-thiophene carboxylic acid as enzyme inhibitors and antibacterial agents.

Solubility
Soluble in ethanol, ether, toluene and chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Tricyclohexyltin chloride
5g
C18H33ClSn
MFCD00003817
acmc-1ck6z
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl
403.622
403.6
123°C to 125°C
3091-32-5
(C6H11)3SnCl
UN3146
OJVZYIKTLISAIH-UHFFFAOYSA-M
chloro(tricyclohexyl)stannane
76539
97%

Chemical Identifiers

3091-32-5
403.622
OJVZYIKTLISAIH-UHFFFAOYSA-M
76539
C1CCC(CC1)[Sn](C2CCCCC2)(C3CCCCC3)Cl
C18H33ClSn
MFCD00003817
acmc-1ck6z
chloro(tricyclohexyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

RTECSNumber : WH6830000

TSCA : TSCA

Documents