Please login to your online account to display your discounted pricing

Alfa Aesar™ Tricyclohexylphosphonium tetrafluoroborate, 99%

$33.46 - $114.66

Chemical Identifiers

Chemical Name or Material Tricyclohexylphosphonium tetrafluoroborate
CAS 58656-04-5
Molecular Formula C18H34P·BF4
Linear Formula (C6H11)3PH€BF4
Formula Weight 368.25
MDL Number MFCD03840580
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2742803

  SDS
alfa aesar™
H2742803
1g Each for $33.46

AAH2742806

  SDS
alfa aesar™
H2742806*
5g Each for $114.66

AAH2742814

  SDS
alfa aesar™
H2742814*
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Tricyclohexylphosphonium tetrafluoroborate
Quantity 1g
CAS 58656-04-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 99%
Molecular Formula C18H34P·BF4
Linear Formula (C6H11)3PH€BF4
Formula Weight 368.25
MDL Number MFCD03840580
Melting Point 166° to 170°C

Applications
Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.