missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tricyclohexylphosphine, technical grade

Manufacturer:  Alfa Aesar™ 4485406

 View more versions of this product

Catalog No. AA4485406



Specifications

Specifications

Tricyclohexylphosphine
Air Sensitive
76°C to 78°C
2622-14-2
MFCD00003853
WLPUWLXVBWGYMZ-UHFFFAOYSA-N
tricyclohexylphosphane
75806
Solid
Unpleasant
White
5g
C18H33P
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
280.436
280.42
Technical

Chemical Identifiers

2622-14-2
280.436
WLPUWLXVBWGYMZ-UHFFFAOYSA-N
75806
C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
C18H33P
MFCD00003853
tricyclohexylphosphine, tricyclohexyl phosphine, phosphine, tricyclohexyl, tricyclohexyl-phosphane, pcy3, tricyclohexylphosphine solution, tchp, p cy 3, phosphorus tricyclohexyl, tricyclohexylphosphine, dissolved in toluene concentration
tricyclohexylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents