Please login to your online account to display your discounted pricing

Alfa Aesar™ Tricyclohexylphosphine oxide

$52.88 - $182.32

Chemical Identifiers

Chemical Name or Material Tricyclohexylphosphine oxide
CAS 13689-19-5
Formula Weight 296.42
MDL Number MFCD00014298
UN Number UN3464
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3038714

  SDS
alfa aesar™
3038714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3038703

  SDS
alfa aesar™
3038703
1g Each for $52.88

AA3038706

  SDS
alfa aesar™
3038706
5g Each for $182.32
Description & Specifications

Specifications

Formula Weight 296.42
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Tricyclohexylphosphine oxide
Physical Form Crystalline
MDL Number MFCD00014298
CAS 13689-19-5
UN Number UN3464
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 155° to 157°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, TOXIC, SOLID, N.O.S.
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.